Potassium bromate 0.1 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

5629-02
JT5629-2CS 615.05 USD
JT5629-2
Potassium bromate 0.1 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Potassium bromate

Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.

Formula: KBrO₃
MW: 167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS Number: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR