Ammonium thiocyanate 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate, Ammonium sulfocyanate

Management of Change (MOC) category = N

5627-02
JT5627-2CS 664.19 USD
JT5627-2
Ammonium thiocyanate 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Ammonium thiocyanate

Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.

Formula: NH₄SCN
MW: 76.12 g/mol
Density: 1.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.095 - 0.105
Standardization at 25°C traceable to NIST Standard Reference Material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR