Cerium(IV) sulfate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ceric sulfate, Cerium disulfate, Cerium disulphate, Ceric sulphate, Cerium(IV) sulfate, Cerium(IV) sulphate

Management of Change (MOC) category = N

5626-02
JT5626-2CS 846.29 USD
JT5626-2
Cerium(IV) sulfate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Cerium (IV) sulphate

Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.

Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8,II
Merck Index: 12,02036

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.095 - 0.105
Standardization at 25°C traceable to NIST Standard Reference Material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR