Bromine 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

5625-02
JT5625-2CS 607.13 USD
JT5625-2
Bromine 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Bromine
Formula: Br₂
MW: 159.81 g/mol
MDL Number: MFCD00010896
CAS Number: 7726-95-6

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.095 - 0.105
Standardization at 25°C traceable to NIST Standard Reference Material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR