Iodine 0.05 mol/l (0.1N), BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

5623-02
JT5623-2CS 2209.78 USD
JT5623-2
Iodine 0.05 mol/l (0.1N), BAKER ANALYZED® volumetric solution, J.T.Baker®
Iodine

BAKER ANALYZED™ Reagent—A grade of high-quality chemicals for laboratory and specialized industrial use.

Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Standardization at 25°C traceable to NIST Standard Reference Material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR