Hydrochloric acid 5 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
Management of Change (MOC) category = NI
5618-07
5618-02
5618-03
JT5618-7EA
413.99
USD
JT5618-7
JT5618-2
JT5618-3
Hydrochloric acid 5 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Hydrochloric acid
The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.
Formula:
HCl MW: 36.46 Density: 1.08 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8 II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Normality | 4.975 - 5.025 |
Sulfate (SO₄) | ≤ 1 ppm |
Ammonium (NH₄) | ≤ 3 ppm |
Trace Impurities - Heavy Metals | ≤ 0.5 ppm |
Trace Impurities - Iron (Fe) | ≤ 0.1 ppm |
Appearance | Passes Test |
Standardization at 25°C traceable to NIST Standard Reference Material. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...