Sulfuric acid 50%, CMOS for the electronics industry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

5253-09
JT5253-9EA 2732.54 USD
JT5253-9
Sulfuric acid 50%, CMOS for the electronics industry, J.T.Baker®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

For Microelectronic Use
Assay (H₂SO₄) 49.0 - 51.0 %
Color (APHA) ≤10
Residue after Ignition ≤3 ppm
Chloride (Cl) ≤1 ppm
Nitrate (NO₃) ≤0.2 ppm
Phosphate (PO₄) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤0.100 ppm
Arsenic and Antimony (as As) ≤0.005 ppm
Trace Impurities - Barium (Ba) ≤1.000 ppm
Trace Impurities - Boron (B) ≤0.010 ppm
Trace Impurities - Cadmium (Cd) ≤0.500 ppm
Trace Impurities - Calcium (Ca) ≤1 ppm
Trace Impurities - Chromium (Cr) ≤0.500 ppm
Trace Impurities - Cobalt (Co) ≤0.500 ppm
Trace Impurities - Copper (Cu) ≤0.010 ppm
Trace Impurities - Gallium (Ga) ≤0.050 ppm
Trace Impurities - Germanium (Ge) ≤0.500 ppm
Trace Impurities - Gold (Au) ≤0.500 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Lead (Pb) ≤0.400 ppm
Trace Impurities - Lithium (Li) ≤1.000 ppm
Trace Impurities - Magnesium (Mg) ≤1.000 ppm
Trace Impurities - Manganese (Mn) ≤0.500 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Trace Impurities - Potassium (K) ≤1.000 ppm
Trace Impurities - Silicon (Si) ≤0.500 ppm
Trace Impurities - Silver (Ag) ≤0.500 ppm
Trace Impurities - Sodium (Na) ≤1.000 ppm
Trace Impurities - Strontium (Sr) ≤0.500 ppm
Trace Impurities - Tin (Sn) ≤0.500 ppm
Trace Impurities - Zinc (Zn) ≤0.500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR