Sodium dihydrogen phosphate monohydrate USP endotoxin tested GMP Manufactured Product

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

4954-60
JT4954-60EA 2288.86 USD
JT4954-60
Sodium dihydrogen phosphate monohydrate USP endotoxin tested GMP Manufactured Product
Sodium dihydrogen phosphate monohydrate
Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100 °C
Density: 2.04 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734

Order Now

Specification Test Results

USP - Assay (anhydrous basis) 98.0 - 103.0 %
USP - Aluminum, Calcium, and Related Elements Passes Test
USP - Arsenic (As) ≤ 3 ppm
USP - Chloride (Cl) ≤ 0.014 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Substances ≤ 0.2 %
USP - pH (1 in 20) 4.1 - 4.5
USP - Sulfate (SO₄) ≤ 0.15 %
USP - Water (H₂O) 10.0 - 15.0 %
Endotoxin Concentration (EU/g ≤ 2.5
Aluminum (Al), For Information Only
Total Aerobic Microbial Coun ≤ 100 CFU/g
Total Yeast and Mold Count ≤ 50 CFU/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR