γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99% (dried basis), purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = NI

4810-09
JT4810-2EA 1383.64 USD
JT4810-2 JT4810-9
γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99% (dried basis), purified, J.T.Baker®
γ-(N-Morpholino)propanesulphonic acid (MOPS)

Suitable for use in biopharmaceutical manufacturing applications.

Formula: C₇H₁₅NO₄S
Melting Pt: 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₇H₁₅NO₄S) (dried basis) ≥99 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
Appearance Passes Test
pH of 1% Solution at 25°C 2.5 - 4.5
Solution (10%) in Water Passes Test
Loss on Drying ≤1 %
Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm ≤0.020
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR