Ammonia solution 20%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

Management of Change (MOC) category = N

4807-05
JT4807-5EA 593.87 USD
JT4807-5
Ammonia solution 20%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Ammonia solution

Certificate of lot analysis provided.

Formula: NH₃
MW: 35.05 g/mol
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (as NH₃) 20 - 22 %
Trace Impurities - Arsenic (As) ≤10 ppt
Trace Impurities - Aluminum (Al) ≤20 ppt
Trace Impurities - Antimony (Sb) ≤10 ppt
Trace Impurities - Barium (Ba) ≤10 ppt
Trace Impurities - Beryllium (Be) ≤10 ppt
Trace Impurities - Bismuth (Bi) ≤10 ppt
Trace Impurities - Cadmium (Cd) ≤10 ppt
Trace Impurities - Calcium (Ca) ≤10 ppt
Trace Impurities - Cerium (Ce) ≤10 ppt
Trace Impurities - Cesium (Cs) ≤10 ppt
Trace Impurities - Chromium (Cr) ≤10 ppt
Trace Impurities - Cobalt (Co) ≤10 ppt
Trace Impurities - Copper (Cu) ≤10 ppt
Trace Impurities - Hafnium (Hf), For Information Only
Trace Impurities - Dysprosium (Dy) ≤10 ppt
Trace Impurities - Erbium (Er) ≤10 ppt
Trace Impurities - Europium (Eu) ≤10 ppt
Trace Impurities - Gadolinium (Gd) ≤10 ppt
Trace Impurities - Iron (Fe) ≤10 ppt
Trace Impurities - Gallium (Ga) ≤10 ppt
Trace Impurities - Germanium (Ge) ≤10 ppt
Trace Impurities - Gold (Au) ≤10 ppt
Trace Impurities - Holmium (Ho) ≤10 ppt
Trace Impurities - Indium (In) ≤10 ppt
Trace Impurities - Lanthanum (La) ≤10 ppt
Trace Impurities - Lead (Pb) ≤10 ppt
Trace Impurities - Lithium (Li) ≤10 ppt
Trace Impurities - Lutetium (Lu) ≤10 ppt
Trace Impurities - Magnesium (Mg) ≤10 ppt
Trace Impurities - Manganese (Mn) ≤10 ppt
Trace Impurities - Mercury (Hg) ≤200 ppt
Trace Impurities - Molybdenum (Mo) ≤10 ppt
Trace Impurities - Neodymium (Nd) ≤10 ppt
Trace Impurities - Nickel (Ni) ≤10 ppt
Trace Impurities - Niobium (Nb) ≤10 ppt
Trace Impurities - Palladium (Pd), For Information Only
Trace Impurities - Platinum (Pt), For Information Only
Trace Impurities - Potassium (K) ≤10 ppt
Trace Impurities - Praseodymium (Pr) ≤10 ppt
Trace Impurities - Rhenium (Re), For Information Only
Trace Impurities - Rhodium (Rh) ≤10 ppt
Trace Impurities - Rubidium (Rb) ≤10 ppt
Trace Impurities - Ruthenium (Ru), For Information Only
Trace Impurities - Samarium (Sm) ≤10 ppt
Trace Impurities - Scandium (Sc) ≤10 ppt
Trace Impurities - Selenium (Se), For Information Only
Trace Impurities - Silver (Ag) ≤10 ppt
Trace Impurities - Sodium (Na) ≤20 ppt
Trace Impurities - Strontium (Sr) ≤10 ppt
Trace Impurities - Terbium (Tb) ≤10 ppt
Trace Impurities - Thallium (Tl) ≤10 ppt
Trace Impurities - Thorium (Th) ≤10 ppt
Trace Impurities - Thulium (Tm) ≤10 ppt
Trace Impurities - Tellurium (Te) ≤10 ppt
Trace Impurities - Tin (Sn) ≤10 ppt
Trace Impurities - Titanium (Ti) ≤10 ppt
Trace Impurities - Tungsten (W) ≤10 ppt
Trace Impurities - Uranium (U) ≤10 ppt
Trace Impurities - Vanadium (V) ≤10 ppt
Trace Impurities - Ytterbium (Yb) ≤10 ppt
Trace Impurities - Yttrium (Y) ≤10 ppt
Trace Impurities - Zinc (Zn) ≤10 ppt
Trace Impurities - Zirconium (Zr) ≤10 ppt
Certificate Provided Reports Actual Lot Analysis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR