Perchloric acid 70%, ULTREX® II, ultrapure Trace_metal, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


M.O.C. Category N

4806-01
JT4806-1EA 1475.71 USD
JT4806-1
Perchloric acid 70%, ULTREX® II, ultrapure Trace_metal, J.T.Baker®
Perchloric acid
Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 39
Melting Pt: -101
Density: 1.60 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (HClO₄) 65 - 71 %
Trace Impurities - Aluminum (Al) ≤100 ppt
Trace Impurities - Antimony (Sb) ≤100 ppt
Trace Impurities - Arsenic (As) ≤100 ppt
Trace Impurities - Barium (Ba) ≤100 ppt
Trace Impurities - Beryllium (Be) ≤100 ppt
Trace Impurities - Bismuth (Bi) ≤10 ppt
Trace Impurities - Cadmium (Cd) ≤10 ppt
Trace Impurities - Calcium (Ca) ≤100 ppt
Trace Impurities - Cesium (Cs) ≤10 ppt
Trace Impurities - Cobalt (Co) ≤100 ppt
Trace Impurities - Copper (Cu) ≤100 ppt
Trace Impurities - Dysprosium (Dy) ≤10 ppt
Trace Impurities - Erbium (Er) ≤10 ppt
Trace Impurities - Europium (Eu) ≤10 ppt
Trace Impurities - Gadolinium (Gd) ≤10 ppt
Trace Impurities - Gallium (Ga) ≤10 ppt
Trace Impurities - Hafnium (Hf), For Information Only
Trace Impurities - Holmium (Ho) ≤10 ppt
Trace Impurities - Indium (In) ≤10 ppt
Trace Impurities - Iron (Fe) ≤100 ppt
Trace Impurities - Lanthanum (La) ≤10 ppt
Trace Impurities - Lead (Pb) ≤10 ppt
Trace Impurities - Lithium (Li) ≤100 ppt
Trace Impurities - Lutetium (Lu) ≤10 ppt
Trace Impurities - Magnesium (Mg) ≤100 ppt
Trace Impurities - Manganese (Mn) ≤100 ppt
Trace Impurities - Molybdenum (Mo) ≤100 ppt
Trace Impurities - Neodymium (Nd) ≤10 ppt
Trace Impurities - Nickel (Ni) ≤100 ppt
Trace Impurities - Niobium (Nb), For Information Only
Trace Impurities - Palladium (Pd) ≤10 ppt
Trace Impurities - Platinum (Pt) ≤100 ppt
Trace Impurities - Potassium (K) ≤100 ppt
Trace Impurities - Praseodymium (Pr) ≤10 ppt
Trace Impurities - Rhodium (Rh) ≤10 ppt
Trace Impurities - Rubidium (Rb) ≤10 ppt
Trace Impurities - Samarium (Sm) ≤10 ppt
Trace Impurities - Scandium (Sc) ≤100 ppt
Trace Impurities - Silver (Ag) ≤100 ppt
Trace Impurities - Sodium (Na) ≤100 ppt
Trace Impurities - Strontium (Sr) ≤100 ppt
Trace Impurities - Tellurium (Te) ≤10 ppt
Trace Impurities - Terbium (Tb) ≤10 ppt
Trace Impurities - Thallium (Tl) ≤100 ppt
Trace Impurities - Thorium (Th) ≤10 ppt
Trace Impurities - Thulium (Tm) ≤10 ppt
Trace Impurities - Tin (Sn) ≤100 ppt
Trace Impurities - Titanium (Ti) ≤100 ppt
Trace Impurities - Tungsten (W), For Information Only
Trace Impurities - Uranium (U) ≤10 ppt
Trace Impurities - Vanadium (V) ≤100 ppt
Trace Impurities - Ytterbium (Yb) ≤10 ppt
Trace Impurities - Yttrium (Y) ≤10 ppt
Trace Impurities - Zinc (Zn) ≤100 ppt
Trace Impurities - Zirconium (Zr) ≤100 ppt
Trace Impurities - Cerium (Ce) ≤10 ppt
Trace Impurities - Tantalum (Ta), For Information Only
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR