Sulfuric acid concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

4700-01
JT4700-1CS 472.31 USD
JT4700-1
Sulfuric acid concentrate for dilution to 1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sulfuric acid

(1 equiv.=49.04 g H2SO4)

Formula: H₂SO₄
MW: 98.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

For laboratory, research or manufacturing use.
Normality (by titrimetry) Passes Test
(Makes 1N solution after dilution to 1000 ml)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR