Zinc sulfate heptahydrate ≥99%, granular USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = R
4383-01
JT4383-1CS
2135.94
USD
JT4383-1
Zinc sulfate heptahydrate ≥99%, granular USP, Multi-Compendial, J.T.Baker®
Zinc sulphate heptahydrate
Formula:
ZnSO₄·7H₂O Melting Pt: 100 °C Density: 1.97 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149894 CAS Number: 7446-20-0 EINECS: 231-793-3 UN: 3077 ADR: 9,III Merck Index: 12,10293 |
Specification Test Results
Meets U.S.P Requirements | |
Meets E.P. Chemical Specifications | |
Meets B.P. Chemical Specifications | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Acidity | Passes Test |
USP - Alkalies and Alkaline Earths | ≤0.9 % |
USP - Arsenic (As) | ≤14 ppm |
USP - Lead (Pb) | ≤0.002 % |
USP - Assay (ZnSO₄·7H₂O) | 99.0 - 108.7 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - pH | 4.4 - 5.6 |
EP/BP - Chloride (Cl) | ≤300 ppm |
EP/BP - Iron (Fe) | ≤100 ppm |
EP/BP - Assay (ZnSO₄·7H₂O) (dried basis) | 99.0 - 104.0 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...