Zinc(II) nitrate hexahydrate 99.0-101.0% (by EDTA titration), BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Zinc nitrate hexahydrate

Management of Change (MOC) category = N

4344-01 4344-05
JT4344-1CS 700.41 USD
JT4344-1 JT4344-5
Zinc(II) nitrate hexahydrate 99.0-101.0% (by EDTA titration), BAKER ANALYZED®
Zinc (II) nitrate hexahydrate

Lot analysis on label.

Formula: ZnN₂O₆·6H₂O
MW: 297.49 g/mol
Boiling Pt: ∼105 °C (1013 hPa)
Melting Pt: 36
Density: 2.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149889
CAS Number: 10196-18-6
EINECS: 231-943-8
UN: 1514
ADR: 5.1 II
Merck Index: 12,10273

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
IMPORTANT: Store below 24°C (75°F). Ship in refrigerated trucks only.
Assay (Zn(NO₃)₂·6H₂O) (by EDTA titrn) 99.0 - 101.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 3.5 - 5.5
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Lead (Pb) ≤0.005 %
Trace Impurities - Copper (Cu) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR