Zinc chloride 97.0-100.5%, granular USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = RL

4320-07
JT4320-7EA 2352.92 USD
JT4320-7
Zinc chloride 97.0-100.5%, granular USP, J.T.Baker®
Zinc chloride

Granular. GMP manufactured. Preserve in a tight container.

Formula: ZnCl₂
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
Store at 2 to 25 °C
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Identification Passes Test
Oxychloride Passes Test
Sulfate (SO₄) ≤0.03 %
Alkalies and Alkaline Earths ≤1.0 %
Ammonium Salts Passes Test
Lead (Pb) ≤0.005 %
Assay (ZnCl₂) 97.0 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR