Zinc ≥99.8% (by EDTA titration), granular, BAKER ANALYZED® ACS (30 Mesh)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

4248-01
JT4248-1CS 2786.47 USD
JT4248-1 JT4248-5
Zinc ≥99.8% (by EDTA titration), granular, BAKER ANALYZED® ACS (30 Mesh)
Zinc

Lot analysis on label.

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Zn) (by EDTA titrn) ≥99.8 %
Iron (Fe) ≤0.01 %
Lead (Pb) ≤0.01 %
Suitability for Arsenic Determination Passes Test
Trace Impurities - Arsenic (As) ≤0.100 ppm
Mesh - on U.S. No. 30 Sieve ≤5 %
Mesh - Thru U.S. No. 50 Sieve ≤30 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR