Urea 99.0-100.5% USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLCSpecification Test Results
GMP Manufactured Product | |
Meets U.S.P Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (NH₂CONH₂) | 98.0 - 102.0 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Residue on Ignition | ≤0.1 % |
USP - Alcohol Insoluble Matter | ≤0.04 % |
USP - Organic impurities: Urea related compound A | ≤0.1 % |
USP - Organic impurities: Any individual unspecified impurity | ≤0.1 % |
USP - Organic impurities: Total impurities | ≤2.0 % |
Solution Test | Passes Test |
Non-Metallic Impurities - Arsenic (As), For Information Only | |
Metallic Impurities - Aluminum (Al), For Information Only | |
Metallic Impurities - Barium (Ba), For Information Only | |
Metallic Impurities - Bismuth (Bi), For Information Only | |
Metallic Impurities - Cadmium (Cd), For Information Only | |
Metallic Impurities - Calcium (Ca), For Information Only | |
Metallic Impurities - Chromium (Cr), For Information Only | |
Metallic Impurities - Cobalt (Co), For Information Only | |
Metallic Impurities - Lead (Pb), For Information Only | |
Metallic Impurities - Lithium (Li), For Information Only | |
Metallic Impurities - Magnesium (Mg), For Information Only | |
Metallic Impurities - Manganese (Mn), For Information Only | |
Metallic Impurities - Mercury (Hg), For Information Only | |
Metallic Impurities - Molybdenum (Mo), For Information Only | |
Metallic Impurities - Nickel (Ni), For Information Only | |
Metallic Impurities - Potassium (K), For Information Only | |
Metallic Impurities - Sodium (Na), For Information Only | |
Metallic Impurities - Strontium (Sr), For Information Only | |
Metallic Impurities - Vanadium (V), For Information Only | |
Metallic Impurities - Zinc (Zn), For Information Only | |
Filtered through a 0.2 micron filter. | |
Residual Solvents: No Class 1,2,3 or other solvents are used or produced | |
in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...