TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

Management of Change (MOC) category = R

4126-07
JT4126-07CS 3021.94 USD
JT4126-07
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Biotech Reagent, J.T.Baker®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
MDL Number: MFCD00012590
CAS Number: 1185-53-1

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NH₂C(CH₂OH)₃·HCl) 99.0 - 101.0 %
Appearance Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Endotoxin Concentration (EU/g) ≤2.5
Identification (by IR) Passes Test
Melting Point 147 - 153 °C
pH of 0.5M Solution at 25°C 3.5 - 5.0
Loss on Drying at 105°C ≤0.5 %
Trace Impurities - Arsenic (As) ≤10 ppm
Trace Impurities - Calcium (Ca) ≤10.0 ppm
Trace Impurities - Copper (Cu) ≤10 ppm
Trace Impurities - Iron (Fe) ≤10 ppm
Trace Impurities - Lead (Pb) ≤10 ppm
Trace Impurities - Magnesium (Mg) ≤10 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR