TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

Management of Change (MOC) category = R

4108-07
JT4108-07CS 4904.54 USD
JT4108-07
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
Formula: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
MDL Number: MFCD00012590
CAS Number: 1185-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (NH₂C(CH₂OH)₃·HCl) (dried basis) ≥99.0 %
Appearance Passes Test
Identification (by IR) Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Trace Impurities - Calcium (Ca) ≤1 ppm
Trace Impurities - Copper (Cu) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤1 ppm
Trace Impurities - Magnesium (Mg) ≤1.000 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by
BioSpectra, Inc., Stroudsburg, PA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR