Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

4095-02 4095-04 4095-05 4095-07
JT4095-2EA 667.42 USD
JT4095-2 JT4095-4 JT4095-5 JT4095-7
Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Sodium dodecyl sulphate (SDS)

For electrophoresis and protein solubilization.


  • Lot analysis on label

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 0.40 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 2926
ADR: 4.1 (6.1),II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (as C₁₂H₂₅OSO₃Na) ≥99 %
Absorbance of a 3% w/v Aqueous Solution at 280 nm (1-cm path) ≤0.1
pH of 1% Solution at 25°C 5.0 - 8.0
Chloride (Cl) ≤0.03 %
Nitrogen (N) ≤5 ppm
Phosphate (PO₄) ≤1 ppm
Heavy Metals (as Pb) ≤2 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR