Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

Management of Change (MOC) category = N

4095-02 4095-04 4095-05 4095-07
JT4095-2EA 667.42 USD
JT4095-2 JT4095-4 JT4095-5 JT4095-7
Sodium dodecyl sulfate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Sodium dodecyl sulphate (SDS)

For electrophoresis and protein solubilization.


  • Lot analysis on label

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 0.40 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 2926
ADR: 4.1 (6.1),II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (as C₁₂H₂₅OSO₃Na) ≥99 %
Absorbance of a 3% w/v Aqueous Solution at 280 nm (1-cm path) ≤0.1
pH of 1% Solution at 25°C 5.0 - 8.0
Chloride (Cl) ≤0.03 %
Nitrogen (N) ≤5 ppm
Phosphate (PO₄) ≤1 ppm
Heavy Metals (as Pb) ≤2 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR