Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for LC-MS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
Management of Change (MOC) category = N
4059-02
4059-06
4059-07
4059-00
JT4059-2EA
217.71
USD
JT4059-2
JT4059-6
JT4059-7
JT4059-0
Glycine ≥99.5% (dried basis), Ultrapure Bioreagent for Electrophoresis, Liquid Chromatography and Molecular Biology, for LC-MS, J.T.Baker®
Glycine
For electrophoresis, liquid chromatography, and molecular biology. Lot analysis on label.
Formula:
NH₂CH₂COOH Boiling Pt: 181 °C (1013 hPa) Melting Pt: 83 °C Density: 1.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (C₂H₅NO₂) (dried basis, by non-aqueous acid- base titration) | ≥99.5 % |
Homogeneity by TLC | Passes Test |
Residue after Ignition | ≤0.10 % |
Loss on Drying at 105°C | ≤0.2 % |
Readily Carbonizable Substances | Passes Test |
Absorbance of a 1M Solution - 280 nm | ≤0.100 |
RNase Activity (None Detected) | Passes Test |
DNase Activity (None Detected) | Passes Test |
Protease Activity (None Detected) | Passes Test |
Arsenic (As) | ≤3 ppm |
ACS - Heavy Metals (as Pb) | ≤5 ppm |
Iron (Fe) | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...