Glycine 98.5-101.5% (dried basis), BAKER ANALYZED®, Biochemical Reagent

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

Management of Change (MOC) category = N

4057-06 4057-02
JT4057-6EA 441.09 USD
JT4057-6 JT4057-2
Glycine 98.5-101.5% (dried basis), BAKER ANALYZED®, Biochemical Reagent
Glycine

Lot analysis on label.

Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₂H₅NO₂) (dried basis, by non-aqueous acid- base titration) 98.5 - 101.5 %
Residue after Ignition ≤0.10 %
Lead (Pb) ≤5.00 ppm
Description Passes Test
Solubility Passes Test
Loss on Drying at 105°C ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR