EDTA disodium salt dihydrate ≥99%, crystals, Ultrapure Bioreagent for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

4040-03 4040-00 4040-04 4040-01 4040-06
JT4040-3EA 136.77 USD
JT4040-3 JT4040-0 JT4040-4 JT4040-1 JT4040-6
EDTA disodium salt dihydrate ≥99%, crystals, Ultrapure Bioreagent for molecular biology, J.T.Baker®
EDTA disodium salt dihydrate

Divalent cation chelating agent


  • Lot analysis on label

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O) 99.0 - 101.0 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
pH of 5% Solution at 25°C 4.0 - 6.0
Calcium (Ca) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Insoluble Matter ≤0.005 %
Iron (Fe) ≤0.005 %
Nitrilotriacetic Acid (by polarography) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR