Sodium acetate trihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = NI

4009-04 4009-06 4009-02
JT4009-4EA 335.97 USD
JT4009-4 JT4009-6 JT4009-2
Sodium acetate trihydrate 99.0-101.0%, crystals, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Sodium acetate trihydrate

Crystals. For liquid chromatography and molecular biology applications. Lot analysis on label.

Formula: H₃CCOONa·3H₂O
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃COONa·3H₂O) 99.0 - 101.0 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
pH of 5% Solution at 25°C 7.5 - 9.2
Absorbance of a 1M Solution - 254 nm ≤0.02
Absorbance of a 1M Solution - 280 nm ≤0.01
Absorbance of a 1M Solution - 350 nm ≤0.01
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
Substances Reducing Permanganate Passes Test
Sulfate (SO₄) ≤0.002 %
Calcium, Magnesium, and R₂O₃ Precipitate ≤0.01 %
Potassium (K) ≤0.005 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR