Tin(II) chloride dihydrate 98.0-103.0%, crystals, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Stannous chloride dihydrate
BAKR3980-01
BAKR3980-05
JT3980-01CS
2746.4
USD
JT3980-01
JT3980-05
Tin(II) chloride dihydrate 98.0-103.0%, crystals, BAKER ANALYZED® ACS
Tin (II) chloride dihydrate
Stannous chloride, dihydrate, crystal suitable for mercury determination.
- Tin(II) chloride, dihydrate
Formula:
SnCl₂·2H₂O Boiling Pt: 623 °C (1013 hPa) Melting Pt: 246 °C Density: 2.71 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149863 CAS Number: 10025-69-1 EINECS: 231-868-0 UN: 3260 ADR: 8,III Merck Index: 13,08861 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (SnCl₂·2H₂O) | 98.0 - 103.0 % |
Solubility in HCl | Passes Test |
Sulfate (SO₄) | Passes Test |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤0.003 % |
Lead (Pb) | ≤0.01 % |
Magnesium (Mg) | ≤0.01 % |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.010 % |
Trace Impurities - Arsenic (As) | ≤2.000 ppm |
Trace Impurities - Mercury (Hg) | ≤0.050 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...