Disodium hydrogen phosphate heptahydrate ≥98% (dried basis), crystals USP suitable for use as excipient, for biochemistry

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Disodium phosphate (heptahydrate), Disodium hydrogenorthophosphate 7-hydrate, Sodium phosphate dibasic 7-hydrate, Sodium monohydrogen phosphate 7-hydrate, Sodium phosphate dibasic heptahydrate, Disodium hydrogenorthophosphate heptahydrate, di-Sodium hydrogen phosphate 7-hydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

Management of Change (MOC) category = HR

3817-01 3817-05 3817-07 3817-QM
JT3817-1CS 702.38 USD
JT3817-1 JT3817-5 JT3817-7 JT3817-QM
Disodium hydrogen phosphate heptahydrate ≥98% (dried basis), crystals USP suitable for use as excipient, for biochemistry
di-Sodium hydrogen phosphate heptahydrate

Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives. All the products are cGMP produced and meet USP grade standards.

Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 231-448-7

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (Na₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤16 ppm
USP - Chloride (Cl) ≤0.06 %
USP - Identification Passes Test
USP - Insoluble Substances ≤0.4 %
USP - Loss on Drying at 130°C 43.0 - 50.0 %
USP - Sulfate (SO₄) ≤0.2 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR