Sodium hydroxide ≥95%, pellets NF, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Management of Change (MOC) category = R

3728-07 3728-28 3728-70
JT3728-7EA 877.98 USD
JT3728-7 JT3728-28 JT3728-20 JT3728-R JT3728-70
Sodium hydroxide ≥95%, pellets NF, FCC, J.T.Baker®
Sodium hydroxide

Pellets. GMP manufactured.


  • Food GMP manufactured product


Storage: PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.

Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Food GMP Manufactured Product
GMP Manufactured Product
Meets N.F. Requirements
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Insoluble Substances and Organic Matter Passes Test
NF - Potassium (K) ≤0.5 %
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
NF - Carbonate (as Na₂CO₃) ≤3.0 %
FCC - Insoluble Substances and Organic Matter Passes Test
FCC - Assay 95.0 - 100.5 %
FCC - Carbonate (as Na₂CO₃) ≤3.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg) ≤0.1 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
STORAGE: Preserve in tight containers.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR