Sodium hydroxide ≥95%, pellets NF, Multi-Compendial for cleaning and processing, for DNA extraction, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Management of Change (MOC) category = R

3718-07 3718-20 3718-15 3718-70
JT3718-7EA 1583.23 USD
JT3718-7 JT3718-20 JT3718-15 JT3718-70
Sodium hydroxide ≥95%, pellets NF, Multi-Compendial for cleaning and processing, for DNA extraction, J.T.Baker®
Sodium hydroxide

Meets N.F. Requirements, Meets J.P. Chemical Specifications, Meets E.P. Chemical Specifications, Meets B.P. Chemical Specifications, GMP Manufactured Product


  • GMP Manufactured Product
  • Bulk Pharmaceutical Chemical


Caution: For Manufacturing, processing or repackaging

Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥ 11.0
NF - Insoluble Substances and Organic Matter Passes Test
NF - Potassium (K) ≤ 0.5 %
NF - Carbonate (as Na₂CO₃) ≤ 3.0 %
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Carbonate (as Na₂CO₃) ≤ 2.0 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 200 ppm
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Assay (NaOH) 97.0 - 100.5 %
JP - Assay 95.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.050 %
JP - Potassium (K) Passes Test
JP - Carbonate (as Na₂CO₃) ≤ 2.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR