tri-Sodium citrate dihydrate ≥99% (by anhydrous basis), granules USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

Management of Change (MOC) category = R

3648-07
JT3648-7EA 1320.41 USD
JT3648-7
tri-Sodium citrate dihydrate ≥99% (by anhydrous basis), granules USP, Multi-Compendial, J.T.Baker®
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
Melting Pt: 150 °C
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - Alkalinity Passes Test
USP - Water (H₂O) 10.0 - 13.0 %
USP - Tartrate Passes Test
USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Readily Carbonizable Substances Passes Test
EP/BP - Chloride (Cl) ≤50 ppm
EP/BP - Oxalate (C₂O₄) ≤300 ppm
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Water (H₂O) 11.0 - 13.0 %
EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 101.0 %
EP/BP - Endotoxin Concentration (EU/g) ≤2.5
JP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis) 99.0 - 101.0 %
JP - pH (1 in 20) 7.5 - 8.5
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.015 %
JP - Identification Passes Test
JP - Sulfate (SO₄) ≤0.048 %
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Tartrate Passes Test
JP - Oxalate (C₂O₄) Passes Test
JP - Readily Carbonizable Substances Passes Test
JP - Loss on Drying at 180°C 10.0 - 13.0 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR