tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granules, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

Management of Change (MOC) category = N

3646-01 3646-07 3646-05
JT3646-1CS 534.17 USD
JT3646-1 JT3646-7 JT3646-R JT3646-5
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granules, BAKER ANALYZED® ACS, J.T.Baker®
tri-Sodium citrate dihydrate

  • Lot analysis on label

Formula: Na₃C₆H₅O₇·2H₂O
Melting Pt: 150 °C
Density: 1.70 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₅Na₃O₇·2H₂O) (by non-aqueous titration) 99.0 - 101.0 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Ammonia (as NH₃) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR