Sodium chloride ≥99% (dried basis), granules USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

Management of Change (MOC) category = R

3627-05 3627-01 3627-20 3627-07 3627-08 3627-60 3627-68 3627-65 3627-F7
JT3627-5CS 1701.01 USD
JT3627-5 JT3627-1 JT3627-20 JT3627-7 JT3627-8 JT3627-60 JT3627-68 JT3627-65 JT3627-F7
Sodium chloride ≥99% (dried basis), granules USP, Multi-Compendial, J.T.Baker®
Sodium chloride

Granular. GMP manufactured product.


  • Management of Change (MOC) category = R
  • GMP manufactured product

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Appearance of Solution Passes Test
USP - Aluminum (Al) ≤0.2 ppm
USP - Arsenic (As) ≤1 ppm
USP - Barium (Ba) Passes Test
USP - Bromide (Br) ≤100 ppm
USP - Ferrocyanide Passes Test
USP - Iodides (I) Passes Test
USP - Iron (Fe) ≤2 ppm
USP - Loss on Drying at 105°C ≤0.5 %
USP - Magnesium and Alkaline Earth Metals (as Ca) ≤100 ppm
USP - Nitrite (NO₂) (Absorbance Value) ≤0.01
USP - Phosphate (PO₄) ≤25 ppm
USP - Potassium (K) ≤500 ppm
USP - Sulfate (SO₄) ≤200 ppm
USP - Assay (NaCl) (dried basis) 99.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Aluminum (Al) ≤0.2 ppm
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Bromide (Br) ≤100 ppm
EP/BP - Ferrocyanide Passes Test
EP/BP - Iodide (I) Passes Test
EP/BP - Nitrite (NO₂) ≤0.01
EP/BP - Phosphate (PO₄) ≤25 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
EP/BP - Arsenic (As) ≤1 ppm
EP/BP - Barium (Ba) Passes Test
EP/BP - Iron (Fe) ≤2 ppm
EP/BP - Magnesium and Alkaline Earth Metals ≤100 ppm
EP/BP - Loss on Drying at 105°C ≤0.5 %
EP/BP - Potassium (K) ≤500 ppm
EP/BP - Assay (NaCl) (dried basis) 99.0 - 100.5 %
EP/BP - Endotoxin Concentration, IU/g ≤2.5
JP - Assay (NaCl) (dried basis) 99.0 - 100.5 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Bromide (Br) Passes Test
JP - Ferrocyanide Passes Test
JP - Iodide (I) Passes Test
JP - Iron (Fe) Passes Test
JP - Magnesium and Alkaline Earth Metals Passes Test
JP - Phosphate (PO₄) Passes Test
JP - Sulfates (SO₄) Passes Test
JP - Heavy Metals (as Pb) ≤3 ppm
JP - Barium (Ba) Passes Test
JP - Arsenic (As) ≤2 ppm
JP - Loss on Drying ≤0.5 %
Total Aerobic Microbial Count, For Information Only
Total Yeasts and Molds Count, For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Must be subjected to further processing during the preparation of
injectable dosage forms
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR