Sodium chloride ≥99% (by argentometric titration), crystals, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

3624-07 3624-19 3624-06 3624-60 3624-05 BAKR3624-01
JT3624-REA 2847.64 USD
JT3624-R JT3624-20 JT3624-7 JT3624-19 JT3624-6 JT3624-60 JT3624-15 JT3624-69 JT3624-01
Sodium chloride ≥99% (by argentometric titration), crystals, BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Sodium chloride

Achieve control over laboratory variables -- minimizing variability and maximizing consistency of results -- by relying on salts in a variety of grades - ACS, BAKER, etc.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.16 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NaCl) (by Ag titrn) ≥99.0 %
pH of 5% Solution at 25°C 5.0 - 9.0
Insoluble Matter ≤0.005 %
Iodide (I) ≤0.002 %
Bromide (Br) ≤0.01 %
Chlorate and Nitrate (as NO₃) ≤0.003 %
ACS - Phosphate (PO₄) ≤5 ppm
Sulfate (SO₄) ≤0.004 %
Barium (Ba) Passes Test
ACS - Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤2 ppm
Calcium (Ca) ≤0.002 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.005 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR