Sodium carbonate, anhydrous, Multi-Compendial

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

3616-09
JT3616-09EA 4375.33 USD
JT3616-09
Sodium carbonate, anhydrous, Multi-Compendial
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
MDL Number: MFCD00003494
CAS Number: 497-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR