Sodium carbonate, anhydrous ≥99.5% (dried basis), powder, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash
Management of Change (MOC) category = N
3602-07
3602-05
3602-R
3602-01
JT3602-7EA
1299.2
USD
JT3602-7
JT3602-5
JT3602-R
JT3602-1
Sodium carbonate, anhydrous ≥99.5% (dried basis), powder, BAKER ANALYZED® ACS, J.T.Baker®
Sodium carbonate
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Density: 2.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 REACH: 01-2119485498-19 Merck Index: 14,08596 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (Na₂CO₃) (dried basis)(by acidimetry) | ≥99.5 % |
Insoluble Matter | ≤0.01 % |
Loss on Heating at 285°C | ≤1.0 % |
Chloride (Cl) | ≤0.001 % |
Phosphate (PO₄) | ≤0.001 % |
Silica (SiO₂) | ≤0.005 % |
Sulfur Compounds (as SO₄) | ≤0.003 % |
Calcium (Ca) | ≤0.03 % |
Magnesium (Mg) | ≤0.005 % |
Potassium (K) | ≤0.005 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...