Sodium carbonate monohydrate ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
3598-09
3598-07
JT3598-9EA
3124.14
USD
JT3598-9
JT3598-7
JT3598-69
Sodium carbonate monohydrate ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium carbonate monohydrate
Formula:
Na₂CO₃·1H₂O Density: 2.25 g/cm³ |
MDL Number:
MFCD00149177 CAS Number: 5968-11-6 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (Na₂CO₃·H₂O) | ≥99.5 % |
ACS - Insoluble Matter | ≤0.01 % |
ACS - Loss on Drying at 150°C | 13.0 - 15.0 % |
ACS - Chloride (Cl) | ≤0.001 % |
ACS - Calcium (Ca) | ≤0.03 % |
ACS - Silica (SiO₂) | ≤0.005 % |
ACS - Sulfur Compounds (as SO₄) | ≤0.004 % |
ACS - Magnesium (Mg) | ≤0.005 % |
ACS - Potassium (K) | ≤0.005 % |
Phosphate (PO₄) | ≤5 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...