Sodium metabisulfite NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite

Management of Change (MOC) category = R

3550-05
JT3550-5CS 1376.78 USD
JT3550-5
Sodium metabisulfite NF, J.T.Baker®
Sodium metabisulphite
Formula: Na₂S₂O₅
MW: 190.11 g/mol
Density: 1.48 g/cm³
MDL Number: MFCD00167602
CAS Number: 7681-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Limit of Chloride ≤0.05 %
NF - Iron (Fe) ≤0.002 %
NF - Limit of Thiosulfate ≤0.05 %
NF - Assay (SO₂) 65.0 - 67.4 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR