Sodium benzoate ≥99% (by anhydrous basis) NF, Multi-Compendial
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Benzoic acid sodium salt
Management of Change (MOC) category = R
3501-05
3501-20
JT3501-5CS
2169.29
USD
JT3501-5
JT3501-20
Sodium benzoate ≥99% (by anhydrous basis) NF, Multi-Compendial
Sodium benzoate
Formula:
C₆H₅COONa Melting Pt: 410…430 °C Density: 1.44 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012463 CAS Number: 532-32-1 EINECS: 208-534-8 REACH: 01-2119460683-35 Merck Index: 13,08654 |
Specification Test Results
GMP Manufactured Product | |
Meets E.P. Chemical Specifications | |
Meets N.F. Requirements | |
Meets B.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Identification C | Passes Test |
NF - Alkalinity | Passes Test |
NF - Water (H₂O) | ≤ 1.5 % |
NF - Assay (C₆H₅COONa) (anhydrous basis) | 99.0 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Acidity or Alkalinity | Passes Test |
EP/BP - Halogenated Compounds (as Cl) - Ionized Chlorine | ≤ 200 ppm |
EP/BP - Halogenated Compounds (as Cl) - Total Chlorine | ≤ 300 ppm |
EP/BP - Loss on Drying | ≤ 2.0 % |
EP/BP - Assay(C₆H₅COONa)(dried basis) | 99.0 - 100.5 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...