Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

3473-05 3473-R
JT3473-5EA 1156.66 USD
JT3473-5 JT3473-R
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Sodium acetate

GMP manufactured.


Caution: For Manufacturing, processing or repackaging.

Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Food GMP Manufactured Product
GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
USP - Calcium and Magnesium Passes Test
USP - Chloride (Cl) ≤0.035 %
USP - Identification Passes Test
USP - Insoluble Matter ≤0.05 %
USP - Loss on Drying at 120°C ≤1.0 %
USP - pH of 3% Solution at 25°C 7.5 - 9.2
USP - Potassium Compounds Passes Test
USP - Residual Acetic Acid, For Information Only
USP - Sulfate (SO₄) ≤0.005 %
FCC - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
FCC - Alkalinity ≤0.2 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying at 120°C ≤1.0 %
FCC - Potassium Compounds Passes Test
The Class 3 solvent, acetic acid, is used to produce this product.
Residual concentration is controlled to <0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR