Sodium acetate, anhydrous ≥99%, purified, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Acetic acid sodium salt

Management of Change (MOC) category = NI

3472-05
JT3472-5CS 2136.97 USD
JT3472-5
Sodium acetate, anhydrous ≥99%, purified, J.T.Baker®
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃COONa) ≥99.0 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 7.0 - 9.2
Heavy Metals (as Pb) ≤0.002 %
Loss on Drying at 120°C ≤1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR