Sodium acetate trihydrate ≥99%, crystals USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Acetic acid sodium salt trihydrate

Management of Change (MOC) category = HR

3462-01 3462-09 3462-05 3462-07 3462-R
JT3462-1EA 121.07 USD
JT3462-1 JT3462-9 JT3462-5 JT3462-7 JT3462-R
Sodium acetate trihydrate ≥99%, crystals USP, FCC, J.T.Baker®
Sodium acetate trihydrate

Crystals.

Formula: H₃CCOONa·3H₂O
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
Preserve in tight containers, store at room temperature.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
USP - Calcium and Magnesium Passes Test
USP - Chloride (Cl) ≤0.035 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Matter ≤0.05 %
USP - Loss on Drying 38.0 - 41.0 %
USP - pH 7.5 - 9.2
USP - Potassium Compounds Passes Test
USP - Sulfate (SO₄) ≤0.005 %
FCC - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
FCC - Alkalinity ≤0.05 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying 38.0 - 41.0 %
FCC - Potassium Compounds Passes Test
No Class 1, 2, 3 or other solvent is likely to be present in the final
product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR