Sodium acetate trihydrate ≥99%, crystals USP, FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Acetic acid sodium salt trihydrate
Management of Change (MOC) category = HR
3462-01
3462-09
3462-05
3462-07
3462-R
JT3462-1EA
121.07
USD
JT3462-1
JT3462-9
JT3462-5
JT3462-7
JT3462-R
Sodium acetate trihydrate ≥99%, crystals USP, FCC, J.T.Baker®
Sodium acetate trihydrate
Crystals.
Formula:
H₃CCOONa·3H₂O Boiling Pt: 120…123 °C (1013 hPa) Melting Pt: 58 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |
|
Specification Test Results
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
Meets U.S.P Requirements | |
Preserve in tight containers, store at room temperature. | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
USP - Assay (CH₃COONa) (dried basis) | 99.0 - 101.0 % |
USP - Calcium and Magnesium | Passes Test |
USP - Chloride (Cl) | ≤0.035 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Insoluble Matter | ≤0.05 % |
USP - Loss on Drying | 38.0 - 41.0 % |
USP - pH | 7.5 - 9.2 |
USP - Potassium Compounds | Passes Test |
USP - Sulfate (SO₄) | ≤0.005 % |
FCC - Assay (CH₃COONa) (dried basis) | 99.0 - 101.0 % |
FCC - Alkalinity | ≤0.05 % |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Loss on Drying | 38.0 - 41.0 % |
FCC - Potassium Compounds | Passes Test |
No Class 1, 2, 3 or other solvent is likely to be present in the final | |
product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...