Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
Management of Change (MOC) category = N
3426-00
3426-01
3426-04
JT3426-0CS
1796.69
USD
JT3426-0
JT3426-1
JT3426-4
Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS
Silver nitrate
Formula:
AgNO₃ Density: 4.35 g/cm³ |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 UN: 1493 ADR: 5.1,II |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (AgNO₃) | ≥99.7 % |
Clarity of Solution | Passes Test |
Free Acid | Passes Test |
Substances Not Precipitated by HCl | ≤0.01 % |
Sulfate (SO₄) | ≤0.002 % |
Lead (Pb) | ≤0.001 % |
Chloride (Cl) | ≤5 ppm |
Trace Impurities - Copper (Cu) | ≤2.000 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...