Silica gel, BAKER ANALYZED® 80-200 Mesh, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = N
3404-01
JT3404-1CS
2345.49
USD
JT3404-1
Silica gel, BAKER ANALYZED® 80-200 Mesh, J.T.Baker®
Silica gel
For column chromatography and scale-up of TLC procedures. Lot analysis on label.
For info :
Particle Size100 - 425 µm
Particle Shape Irregular
Pore Size 60 Å
Formula:
SiO₂ MW: 60.08 g/mol Boiling Pt: 2230 Melting Pt: 1710 Density: 2.10 kg/m³ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 REACH: 01-2119379499-16 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Loss on Ignition | ≤13 % |
Alcohol-Ether Soluble Substances | ≤0.05 % |
Flow Rate, mL/min. | ≥0.5 |
Mesh - Thru U.S. No. 80 Sieve | ≥90.0 % |
Mesh - Thru U.S. No. 200 Sieve | ≤10.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...