Silica gel, BAKER ANALYZED® (4-10 Mesh) desiccant, with moisture indicator, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
4–10 mesh. Indicating. Lot analysis on label.
Color Change:
Blue.................Activated
Violet..........10% Moisture Absorbed
Pink............19% Moisture Absorbed
Pale Pink.......28% Moisture Absorbed
Formula:
SiO₂ Density: 2.10 kg/m³ |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Loss on Heating at 200°C | ≤6.0 % |
Suitability for Moisture Absorption at 80% Relative Humidity | ≥33 % |
Suitability for Moisture Absorption at 20% Relative Humidity | ≥10 % |
Suitability as Indicating Desiccant | Passes Test |
Mesh - Thru U.S. No. 10 Sieve | ≤5.0 % |
Color Change: Blue.................Activated Violet..........10% Moisture Absorbed Pink............19% Moisture Absorbed Pale Pink.......28% Moisture Absorbed |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...