Sand, quartz, purified, washed and ignited, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

3382-01 3382-05
JT3382-1CS 435.54 USD
JT3382-1 JT3382-5
Sand, quartz, purified, washed and ignited, J.T.Baker®
Sand, quartz

Washed and ignited. For boats.

Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1427 °C
Density: 2.622…2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD02100519
CAS Number: 14808-60-7
EINECS: 238-878-4

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Substances Soluble in HCl ≤0.16 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR