di-Potassium hydrogen phosphate USP, Multi-Compendial (Pyrophosphate Tested)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
3257-65
JT3257-65EA
4729.61
USD
JT3257-65
di-Potassium hydrogen phosphate USP, Multi-Compendial (Pyrophosphate Tested)
di-Potassium hydrogen phosphate
Formula:
K₂HPO₄ MW: 174.18 g/mol |
MDL Number:
MFCD00011383 CAS Number: 7758-11-4 |
Specification Test Results
USP - Assay (K₂HPO₄) (dried basis) | 98.0 - 100.5 % |
USP - Arsenic (As) | ≤ 3 ppm |
USP - Carbonate (CO₃) | Passes Test |
USP - Chloride (Cl) | ≤ 0.03 % |
USP - Endotoxin Concentration (2.5 EU/g) | Passes Test |
USP - Fluoride (F) | ≤ 0.001 % |
USP - Identification A (USP) | Passes Test |
USP - Identification B (USP) | Passes Test |
USP - Insoluble Substances | ≤ 0.2 % |
USP - Iron (Fe) | ≤ 0.003 % |
USP - Loss on Drying | ≤ 1.0 % |
USP - Mono- or Tri- Basic Salt | Passes Test |
USP - pH (1 in 20) | 8.5 - 9.6 |
USP - Sodium (Na) | Passes Test |
USP - Sulfate (SO₄) | ≤ 0.1 % |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Assay (K₂HPO₄) (dried basis) | 98.0 - 101.0 % |
EP/BP - Chloride (Cl) | ≤ 200 ppm |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Iron (Fe) | ≤ 10 ppm |
EP/BP - Loss on Drying | ≤ 2.0 % |
EP/BP - Monopotassium phosphate | ≤ 0.025 |
EP/BP - Reducing Substances | Passes Test |
EP/BP - Sodium (Na) | ≤ 0.1 % |
EP/BP - Sulfate (SO₄) | ≤ 0.1 % |
Color (APHA), For Information Only |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...