Potassium nitrate 99.0-100.5%, granular USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Nitrate of Potash

Management of Change (MOC) category = HR

3192-88
JT3192-88CS 1477.59 USD
JT3192-88
Potassium nitrate 99.0-100.5%, granular USP, FCC, J.T.Baker®
Potassium nitrate
Formula: KNO₃
MW: 101.1 g/mol
Density: 2.10 g/cm³
MDL Number: MFCD00011409
CAS Number: 7757-79-1
UN: 1486
ADR: 5.1,III

Order Now

Specification Test Results

Food GMP Manufactured Product
GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (KNO₃) 99.0 - 100.5 %
USP - Arsenic (As) ≤3 ppm
USP - Chloride (Cl) ≤0.03 %
USP - Identification Passes Test
USP - Iron (Fe) ≤0.001 %
USP - Lead (Pb) (USP) ≤0.001 %
USP - Nitrite (NO₂) ≤5 ppm
USP - Sodium (Na) ≤0.1 %
USP - Sulfate (SO₄) ≤0.1 %
FCC - Assay (KNO₃) (dried basis) 99.0 - 100.5 %
FCC - Chlorate Passes Test
FCC - Identification Passes Test
FCC - Lead (Pb) (FCC) ≤4 mg/kg
FCC - Loss on Drying at 105°C ≤1 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR