Potassium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = NI

3190-05 3190-80
JT3190-5CS 3131.86 USD
JT3190-5 JT3190-1 JT3190-80 JT3190-7
Potassium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KNO₃) ≥99.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 4.5 - 8.5
Chloride (Cl) ≤0.002 %
ACS- Nitrite (NO₂) ≤0.001 %
ACS - Sulfate (SO₄) ≤0.003 %
Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.005 %
Iodate (IO₃) ≤5 ppm
Phosphate (PO₄) ≤3 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR