Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

3104-01 3104-05 3104-07
JT3104-1CS 2053.45 USD
JT3104-1 JT3104-5 JT3104-7
Potassium hexacyanoferrate(III) ≥99.0% (by iodometry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium hexacyanoferrate (III)

  • Lot analysis on label

Formula: K₃Fe(CN)₆
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (K₃Fe(CN)₆) (by iodometry) ≥99.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.01 %
Sulfate (SO₄) ≤0.01 %
Ferro Compounds (as [Fe(CN)₆]⁴⁻) ≤0.05 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR